Assay Details
Target Gene Details
Entrez Gene ID: | 73836 |
Gene Name: | solute carrier family 35, member B2 |
Gene Aliases: |
1110003M08Rik, AA407792, AI842275, PAPST1, Slc35b1 |
Location: |
Chr.17:45563879-45567671 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35b2 | NM_028662.2 | 3 | 1684 | NP_082938.1 |
| AF414190.1 | 4 | 1357 | AAL07487.1 | |
| AK090329.1 | 3 | 1873 | BAC41173.1 | |
| AK144403.1 | 4 | 1380 | BAE25873.1 | |
| AK152362.1 | 4 | 1358 | BAE31152.1 | |
| AK154868.1 | 4 | 1343 | BAE32889.1 | |
| AK155551.1 | 4 | 1390 | BAE33321.1 | |
| AK167343.1 | 3 | 1685 | BAE39443.1 | |
| AK183518.1 | ||||
| AK189197.1 | ||||
| AK198521.1 | ||||
| AK207266.1 | ||||
| AK208869.1 | ||||
| AK211181.1 | ||||
| AK212100.1 | ||||
| AK215217.1 | ||||
| AK215246.1 | ||||
| BC025875.1 | 4 | 1356 | AAH25875.1 | |
| BC036992.1 | 4 | 1364 | AAH36992.1 |
More Information
Set Membership: |
|

Genomic Map