Assay Details
Target Gene Details
Entrez Gene ID: | 224674 |
Gene Name: | solute carrier family 37 (glycerol-3-phosphate transporter), member 1 |
Gene Aliases: |
G3PP |
Location: |
Chr.17:31294798-31350709 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc37a1 | NM_001242427.1 | 20 | 2745 | NP_001229356.1 |
| NM_153062.2 | 20 | 2325 | NP_694702.1 | |
| AK033656.1 | 20 | 2214 | BAC28411.1 | |
| AK143636.1 | 20 | 2751 | BAE25474.1 | |
| AK153706.1 | 20 | 2748 | BAE32150.1 | |
| AK163990.1 | 19 | 1928 | BAE37574.1 | |
| BC027294.1 | 20 | 2325 | AAH27294.1 | |
| BC029208.1 | 12 | 1145 | AAH29208.1 | |
| CO039406.1 | 1 | 623 |
More Information
Set Membership: |
|

Genomic Map