Assay Details
Target Gene Details
Entrez Gene ID: | 20541 |
Gene Name: | solute carrier family 8 (sodium/calcium exchanger), member 1 |
Gene Aliases: |
AI852629, AV344025, D930008O12Rik, Ncx1 |
Location: |
Chr.17:81373105-81738387 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc8a1 | NM_001112798.2 | 2 | 1000 | NP_001106269.1 |
| NM_001286684.1 | 2 | 1000 | NP_001273613.1 | |
| NM_011406.3 | 2 | 1000 | NP_035536.2 | |
| NR_104580.1 | 2 | 1000 | ||
| AB080744.1 | 1 | 69 | BAB85833.1 | |
| AF004666.1 | 2 | 995 | AAB69167.1 | |
| BB660129.1 | ||||
| BC079673.1 | 1 | 891 | AAH79673.1 | |
| DQ388998.1 | 1 | 890 | ABD46717.1 | |
| U70033.1 | 2 | 990 | AAB46708.1 |
More Information
Set Membership: |
|

Genomic Map