Assay Details
Target Gene Details
Entrez Gene ID: | 65962 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2 |
Gene Aliases: |
0610011L07Rik, 1200011K07Rik, 2010007A20Rik, E3karp, NHERF-2, Nherf2, Octs2, Sip-1, Sip1, Sryip1, Tka-1 |
Location: |
Chr.17:24639281-24650340 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc9a3r2 | NM_023055.2 | 1 | 186 | NP_075542.2 |
AB026489.1 | 1 | 159 | BAA97568.1 | |
AK004710.1 | 1 | 182 | ||
AK039491.1 | 1 | 174 | BAC30366.1 | |
AK075800.1 | 1 | 187 | ||
AK075813.1 | 1 | 187 | BAC35980.1 | |
AK084801.1 | 1 | 187 | BAC39282.1 | |
AK169007.1 | 1 | 176 | ||
AK170057.1 | 1 | 172 | BAE41537.1 | |
BC070458.1 | 1 | 170 | AAH70458.1 |
