Assay Details
Target Gene Details
Entrez Gene ID: | 16891 |
Gene Name: | lipase, endothelial |
Gene Aliases: |
3110013K01Rik, EL, lipase, mEDL |
Location: |
Chr.18:74939322-74961263 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lipg | NM_010720.3 | 10 | 2112 | NP_034850.3 |
| AF118768.1 | 10 | 2097 | AAD30435.1 | |
| AK090224.1 | 10 | 2112 | ||
| AK150054.1 | 3 | 495 | BAE29270.1 | |
| AK150061.1 | 3 | 495 | BAE29275.1 | |
| AK152430.1 | 10 | 2110 | BAE31212.1 | |
| AK172101.1 | 10 | 2112 | BAE42826.1 | |
| BC020991.1 | 10 | 1966 | AAH20991.1 |
More Information
Set Membership: |
|

Genomic Map