Assay Details
Target Gene Details
Entrez Gene ID: | 27426 |
Gene Name: | N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase |
Gene Aliases: |
AI596180, UCE |
Location: |
Chr.16:5195280-5204040 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nagpa | NM_013796.3 | 10 | 1769 | NP_038824.2 |
NR_027804.1 | 9 | 1866 | ||
AK032158.1 | 10 | 1770 | BAC27731.1 | |
AK087468.1 | 10 | 1772 | ||
AK138035.1 | 10 | 1762 | BAE23539.1 | |
AK159629.1 | 10 | 1748 | BAE35243.1 | |
AK162633.1 | 10 | 1752 | ||
AK168830.1 | 9 | 1854 | ||
AK170219.1 | 10 | 1751 | BAE41643.1 | |
AK177662.1 | ||||
BC039790.1 | 10 | 1761 | AAH39790.1 |
