Assay Details
Target Gene Details
Entrez Gene ID: | 27426 |
Gene Name: | N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase |
Gene Aliases: |
AI596180, UCE |
Location: |
Chr.16:5195280-5204040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nagpa | NM_013796.3 | 2 | 498 | NP_038824.2 |
NR_027804.1 | 1 | 595 | ||
AK032158.1 | 2 | 499 | BAC27731.1 | |
AK087468.1 | 2 | 498 | ||
AK138035.1 | 2 | 491 | BAE23539.1 | |
AK155450.1 | 2 | 477 | BAE33269.1 | |
AK159629.1 | 2 | 477 | BAE35243.1 | |
AK162633.1 | 2 | 480 | ||
AK168830.1 | 1 | 585 | ||
AK169539.1 | 1 | 574 | ||
AK170219.1 | 2 | 480 | BAE41643.1 | |
AK211874.1 | ||||
BC039790.1 | 2 | 490 | AAH39790.1 |
