Assay Details
Target Gene Details
Entrez Gene ID: | 12211 |
Gene Name: | baculoviral IAP repeat-containing 6 |
Gene Aliases: |
A430032G04Rik, A430040A19Rik, AA501170, Bruce, D630005A10Rik, mKIAA1289 |
Location: |
Chr.17:74528216-74703356 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Birc6 | NM_007566.3 | 7 | 1473 | NP_031592.3 |
AK045446.1 | 7 | 1242 | BAC32373.1 | |
AK079995.1 | 7 | 1250 | BAC37801.1 | |
Y17267.1 | 7 | 1474 | CAA76720.1 |
