Assay Details
Target Gene Details
Entrez Gene ID: | 224997 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 1 |
Gene Aliases: |
4933422O14Rik, 9630002F18, AI845682, AI848168, BB075781, D17Bwg0511e, GKAP/SAPAP, GKPA/SAPAP, Gkap, Sapap1, mKIAA4162 |
Location: |
Chr.17:69969078-70821413 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap1 | NM_001128180.1 | 10 | 2473 | NP_001121652.1 |
| NM_177639.6 | 12 | 3490 | NP_808307.2 | |
| AK016873.1 | 3 | 916 | ||
| AK147448.1 | 15 | 4277 | BAE27919.1 | |
| AK161941.1 | 10 | 2448 | BAE36644.1 | |
| AK220515.1 | 14 | 4340 | BAD90519.1 | |
| BC062120.1 | 12 | 3491 | AAH62120.1 |
More Information
Set Membership: |
|

Genomic Map