Assay Details
Target Gene Details
Entrez Gene ID: | 320624 |
Gene Name: | RIKEN cDNA 4930474G06 gene |
Gene Aliases: |
- |
Location: |
Chr.18:28560164-28998176 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930474G06Rik | NR_045398.1 |

Genomic Map