Assay Details
Target Gene Details
Entrez Gene ID: | 402767 |
Gene Name: | RIKEN cDNA A830052D11 gene |
Gene Aliases: |
- |
Location: |
Chr.18:32359057-32378284 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A830052D11Rik | NR_045403.1 | |||
| AK043919.1 |
Target Gene Details
Entrez Gene ID: | 30948 |
Gene Name: | bridging integrator 1 |
Gene Aliases: |
ALP-1, Amphl, BRAMP-2, SH3P9 |
Location: |
Chr.18:32377217-32435740 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bin1 | NM_001083334.1 | 1 | 222 | NP_001076803.1 |
| NM_009668.2 | 1 | 222 | NP_033798.1 | |
| BC065160.1 | 1 | 194 | AAH65160.1 | |
| BQ573238.1 | 1 | 194 | ||
| CJ121020.1 | 1 | 224 | ||
| U60884.1 | 1 | 210 | AAC52661.1 |
More Information
Set Membership: |
|

Genomic Map