Assay Details
Target Gene Details
Entrez Gene ID: | 30948 |
Gene Name: | bridging integrator 1 |
Gene Aliases: |
ALP-1, Amphl, BRAMP-2, SH3P9 |
Location: |
Chr.18:32377217-32435740 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bin1 | NM_001083334.1 | 15 | 1821 | NP_001076803.1 |
NM_009668.2 | 19 | 2154 | NP_033798.1 | |
AI840452.1 | 1 | 315 | ||
AW496330.1 | 1 | 290 | ||
BC065160.1 | 15 | 1793 | AAH65160.1 |
