Assay Details
Target Gene Details
Entrez Gene ID: | 17968 |
Gene Name: | neural cell adhesion molecule 2 |
Gene Aliases: |
Ncam-2, Ocam, RNCAM |
Location: |
Chr.16:81200697-81624290 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncam2 | NM_001113208.1 | NP_001106679.1 | ||
NM_010954.4 | 16 | 3163 | NP_035084.1 | |
AF001286.1 | 16 | 3089 | AAB69124.1 | |
AF001287.1 | AAB69125.1 | |||
AF016619.1 | 16 | 3045 | AAC53375.1 | |
AF016620.1 | AAC53376.1 |
