Assay Details
Target Gene Details
Entrez Gene ID: | 353130 |
Gene Name: | protease, serine 33 |
Gene Aliases: |
Eos, mT6, tryptase-6 |
Location: |
Chr.17:23833338-23836126 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss33 | NM_001081399.2 | 4 | 320 | NP_001074868.2 |
| AY262280.1 | 4 | 296 | AAP20885.1 | |
| BC101950.1 | 4 | 296 | AAI01951.1 | |
| BC103541.1 | 5 | 392 | AAI03542.1 | |
| BC103542.1 | 4 | 296 | AAI03543.1 | |
| BQ893522.1 | 3 | 720 |
More Information
Set Membership: |
|

Genomic Map