Assay Details
Target Gene Details
Entrez Gene ID: | 63955 |
Gene Name: | CDK5 and Abl enzyme substrate 1 |
Gene Aliases: |
Cables, ik3-1 |
Location: |
Chr.18:11839274-11945627 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cables1 | NM_001146287.1 | 10 | 3147 | NP_001139759.1 |
NM_022021.2 | 9 | 3069 | NP_071304.2 | |
AF133208.1 | 9 | 3068 | AAG33933.1 | |
AK156273.1 | 10 | 3203 | BAE33652.1 | |
CA751142.1 | 1 | 437 | ||
CB205636.1 |
