Assay Details
Target Gene Details
Entrez Gene ID: | 225288 |
Gene Name: | formin homology 2 domain containing 3 |
Gene Aliases: |
A930009H06Rik, FHOS2, mKIAA1695 |
Location: |
Chr.18:24708456-25133507 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhod3 | NM_001289654.1 | 25 | 5482 | NP_001276583.1 |
| NM_001289655.1 | 26 | 5839 | NP_001276584.1 | |
| NM_175276.4 | 27 | 5935 | NP_780485.2 | |
| AB078608.1 | 27 | 5113 | BAC98302.1 | |
| AB078609.1 | 25 | 4660 | BAC98303.1 | |
| AB100088.1 | 26 | 5017 | BAC98348.1 | |
| BC060607.1 | 12 | 2891 | AAH60607.1 | |
| BQ176384.1 | 1 | 317 |
Target Gene Details
Entrez Gene ID: | 66648 |
Gene Name: | tubulin polyglutamylase complex subunit 2 |
Gene Aliases: |
5730437P09Rik, 5730494M16Rik, AI666318, Pgs2 |
Location: |
Chr.18:25127223-25169007 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpgs2 | NM_001142698.1 | NP_001136170.1 | ||
| AK141632.1 |
More Information
Set Membership: |
|

Genomic Map