Assay Details
Target Gene Details
Entrez Gene ID: | 80859 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta |
Gene Aliases: |
AA408868, INAP, Mail |
Location: |
Chr.16:55811377-55838641 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbiz | NM_001159394.1 | 12 | 2682 | NP_001152866.1 |
| NM_001159395.1 | 13 | 2564 | NP_001152867.1 | |
| NM_030612.3 | 14 | 2912 | NP_085115.1 | |
| AB020974.1 | 14 | 3124 | BAB18302.1 | |
| AK082908.1 | 12 | 2681 | BAC38681.1 | |
| AK136509.1 | 9 | 2151 | BAE23017.1 | |
| AK150519.1 | 12 | 2681 | BAE29631.1 | |
| AK156826.1 | 12 | 2682 | ||
| AK159171.1 | 12 | 2683 | BAE34871.1 | |
| AK159459.1 | 12 | 2683 | BAE35101.1 | |
| AK159917.1 | 12 | 2683 | BAE35480.1 |
More Information
Set Membership: |
|

Genomic Map