Assay Details
Target Gene Details
Entrez Gene ID: | 52633 |
Gene Name: | nitrilase family, member 2 |
Gene Aliases: |
1190017B19Rik, D16Ertd502e |
Location: |
Chr.16:57156665-57167332 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nit2 | NM_023175.1 | 10 | 1122 | NP_075664.1 |
| AF284573.1 | 10 | 1122 | AAF87102.1 | |
| AK052922.1 | 1 | 1053 |
More Information
Set Membership: |
|

Genomic Map