Assay Details
Target Gene Details
Entrez Gene ID: | 67824 |
Gene Name: | NmrA-like family domain containing 1 |
Gene Aliases: |
1110025F24Rik, AI256624 |
Location: |
Chr.16:4711317-4719356 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nmral1 | NM_001290761.1 | 5 | 1248 | NP_001277690.1 |
NM_001290762.1 | 5 | 1325 | NP_001277691.1 | |
NM_026393.2 | 6 | 1278 | NP_080669.1 | |
AA166262.1 | 1 | 315 | ||
AK012340.1 | 5 | 1081 | BAB28172.1 | |
BC030039.1 | 6 | 981 | AAH30039.1 |
