Assay Details
Target Gene Details
Entrez Gene ID: | 14724 |
Gene Name: | glycoprotein Ib, beta polypeptide |
Gene Aliases: |
- |
Location: |
Chr.16:18620319-18622403 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gp1bb | NM_001001999.1 | 1 | 433 | NP_001001999.1 |
| NM_010327.2 | 1 | 433 | NP_034457.1 | |
| BC138156.1 | 1 | 387 | ||
| BC145160.1 | 1 | 387 |
Target Gene Details
Entrez Gene ID: | 18951 |
Gene Name: | septin 5 |
Gene Aliases: |
Cdcrel-1, Cdcrel1, Pnutl1 |
Location: |
Chr.16:18621811-18629938 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept5 | NM_213614.2 | 12 | 1967 | NP_998779.2 |
| AF033350.1 | 10 | 1814 | AAC83974.1 | |
| AK134700.1 | 12 | 1904 | BAE22248.1 | |
| AK186465.1 | ||||
| BC059848.1 | 10 | 1845 | AAH59848.1 | |
| BC079631.1 | 6 | 1281 | AAH79631.1 | |
| BC094347.1 | 11 | 1843 | AAH94347.1 |
More Information
Set Membership: |
|

Genomic Map