Assay Details
Target Gene Details
Entrez Gene ID: | 18951 |
Gene Name: | septin 5 |
Gene Aliases: |
Cdcrel-1, Cdcrel1, Pnutl1 |
Location: |
Chr.16:18621811-18629938 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept5 | NM_213614.2 | 12 | 1302 | NP_998779.2 |
| AF033350.1 | 10 | 1149 | AAC83974.1 | |
| AK134700.1 | 12 | 1239 | BAE22248.1 | |
| BC059848.1 | 10 | 1180 | AAH59848.1 | |
| BC079631.1 | 6 | 616 | AAH79631.1 | |
| BC094347.1 | 11 | 1178 | AAH94347.1 | |
| BC141073.1 | ||||
| BC145331.1 | ||||
| BC145332.1 |
More Information
Set Membership: |
|

Genomic Map