Assay Details
Target Gene Details
Entrez Gene ID: | 106369 |
Gene Name: | yippee-like 1 (Drosophila) |
Gene Aliases: |
0610009L05Rik, 1700016N17Rik, 1700019O22Rik, 4921520K19Rik, 4930511F14Rik, AV118478, Dgl1, Ppil2, mdgl-1 |
Location: |
Chr.16:17070138-17086736 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ypel1 | NM_001291047.1 | 6 | 2226 | NP_001277976.1 |
NM_001291053.1 | 4 | 1820 | NP_001277982.1 | |
NM_023249.6 | 5 | 2100 | NP_075738.1 | |
AF190624.1 | 5 | 2016 | AAG17143.1 | |
AK006137.1 | 4 | 1718 | BAC25133.1 | |
AK014931.1 | 5 | 1926 | BAB29625.1 | |
AK133401.1 | BAE21635.1 | |||
AK139453.1 | 5 | 2008 | BAE24018.1 |
