Assay Details
Target Gene Details
Entrez Gene ID: | 268973 |
Gene Name: | NLR family, CARD domain containing 4 |
Gene Aliases: |
9530011P19Rik, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, Card12, IPAF |
Location: |
Chr.17:74425716-74459143 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrc4 | NM_001033367.3 | 4 | 1807 | NP_001028539.1 |
| AK035300.1 | 1 | 1208 | ||
| AK143864.1 | 4 | 1808 | BAE25573.1 | |
| AK171624.1 | 4 | 1802 | BAE42570.1 |

Genomic Map