Assay Details
Target Gene Details
Entrez Gene ID: | 94224 |
Gene Name: | steroid 5 alpha-reductase 2 |
Gene Aliases: |
5ART2, S5AR 2 |
Location: |
Chr.17:74017706-74047916 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srd5a2 | NM_053188.2 | 1 | 196 | NP_444418.1 |
| AB049456.1 | 1 | 196 | BAB40179.1 | |
| AK138946.1 | 1 | 137 | BAE23830.1 | |
| BC125510.1 | 1 | 180 |
More Information
Set Membership: |
|

Genomic Map