Assay Details
Target Gene Details
Entrez Gene ID: | 224829 |
Gene Name: | transcriptional regulating factor 1 |
Gene Aliases: |
9430096I18Rik, AI429294, B830015H24, RAPA, Trep-132, Trep132 |
Location: |
Chr.17:47140942-47359458 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trerf1 | NM_001097623.1 | 6 | 1218 | NP_001091092.1 |
NM_172622.2 | 3 | 1055 | NP_766210.1 | |
AK035176.1 | 1 | 824 | ||
AK042320.1 | 4 | 1359 | BAC31222.1 | |
AK046822.1 | 1 | 697 | BAC32884.1 | |
AK154542.1 | 5 | 1230 | ||
AK155049.1 | 5 | 1332 | ||
AK155404.1 | 5 | 1231 | ||
AK170342.1 | 5 | 1326 | BAE41732.1 | |
AY081774.1 | 2 | 968 | AAN02288.1 | |
BC059215.1 | 6 | 1218 | AAH59215.1 | |
BC108416.1 | 1 | 678 | AAI08417.1 |
