Assay Details
Target Gene Details
Entrez Gene ID: | 79263 |
Gene Name: | tripartite motif-containing 39 |
Gene Aliases: |
1100001D15Rik, E130103K13Rik, RBCC-B30.2, Rnf23, mKIAA4179, tfp |
Location: |
Chr.17:36258872-36272004 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim39 | NM_024468.2 | 7 | 2065 | NP_077788.2 |
| NM_178281.1 | 7 | 2050 | NP_840065.1 | |
| AB046382.1 | 7 | 2065 | BAB16375.1 | |
| AK003155.1 | 1 | 122 | ||
| AK145861.1 | 7 | 2003 | ||
| AK166148.1 | 7 | 1954 | BAE38597.1 | |
| AK220298.1 | 7 | 1986 | BAD90223.1 | |
| BC031540.1 | 6 | 2131 | AAH31540.1 |
More Information
Set Membership: |
|

Genomic Map