Assay Details
Target Gene Details
Entrez Gene ID: | 22436 |
Gene Name: | xanthine dehydrogenase |
Gene Aliases: |
XO, Xor, Xox-1, Xox1 |
Location: |
Chr.17:73883895-73950196 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xdh | NM_011723.3 | 36 | 4406 | NP_035853.2 |
| AI323529.1 | 1 | 218 | ||
| AK008517.1 | BAB25715.1 | |||
| AK036869.1 | ||||
| AK090058.1 | 27 | 3549 | ||
| AK134156.1 | 22 | 2892 | ||
| AK143412.1 | 36 | 4357 | ||
| AK144700.1 | 36 | 4358 | BAE26022.1 | |
| AK160010.1 | 33 | 4105 | ||
| AK161529.1 | 36 | 7582 | ||
| BC003997.1 | 5 | 878 | AAH03997.1 | |
| X62932.1 | 36 | 4340 | CAA44705.1 |
More Information
Set Membership: |
|

Genomic Map