Assay Details
Target Gene Details
Entrez Gene ID: | 106338 |
Gene Name: | NOL1/NOP2/Sun domain family member 3 |
Gene Aliases: |
6720484A09Rik, AU022521 |
Location: |
Chr.16:62732444-62786794 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun3 | NM_178925.3 | 3 | 388 | NP_849256.1 |
| AK032127.1 | 3 | 430 | BAC27717.1 | |
| AK084698.1 | 3 | 382 | ||
| AK153942.1 | BAE32268.1 | |||
| AK169841.1 | BAE41405.1 | |||
| BC125628.1 | 3 | 388 | ||
| BC125630.1 | 3 | 388 |
More Information
Set Membership: |
|

Genomic Map