Assay Details
Target Gene Details
Entrez Gene ID: | 224020 |
Gene Name: | phosphatidylinositol 4-kinase, catalytic, alpha polypeptide |
Gene Aliases: |
Pik4ca |
Location: |
Chr.16:17280351-17406314 on Build GRCm38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pi4ka | NM_001001983.2 | NP_001001983.2 | ||
| BC075629.1 | AAH75629.1 |
Target Gene Details
Entrez Gene ID: | 15160 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade D, member 1 |
Gene Aliases: |
AA985900, AI303446, HCII, Hcf2 |
Location: |
Chr.16:17331339-17343574 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpind1 | NM_008223.3 | 3 | 1097 | NP_032249.3 |
| BC034543.1 | 2 | 904 | AAH34543.1 | |
| BC089610.1 | 3 | 1053 | AAH89610.1 | |
| CB949626.1 | 2 | 497 | ||
| U07425.1 | 2 | 964 | AAA18452.1 |
More Information
Set Membership: |
|

Genomic Map