Assay Details
Target Gene Details
Entrez Gene ID: | 13640 |
Gene Name: | ephrin A5 |
Gene Aliases: |
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS |
Location: |
Chr.17:62602957-62881317 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efna5 | NM_010109.3 | 1 | 156 | NP_034239.1 |
| NM_207654.2 | 1 | 156 | NP_997537.1 | |
| BC040218.1 | 1 | 156 | AAH40218.1 | |
| U90664.1 | AAB50239.1 | |||
| U90665.1 | AAB50240.1 |
More Information
Set Membership: |
|

Genomic Map