Assay Details
Target Gene Details
Entrez Gene ID: | 106581 |
Gene Name: | integrin alpha FG-GAP repeat containing 3 |
Gene Aliases: |
- |
Location: |
Chr.17:26212692-26244242 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itfg3 | NM_001206335.1 | 13 | 2180 | NP_001193264.1 |
| NM_207217.4 | 12 | 2068 | NP_997100.1 | |
| AK029427.1 | 12 | 1968 | BAC26447.1 | |
| AK170646.1 | 13 | 2099 | BAE41933.1 | |
| AK184216.1 | ||||
| BC046457.1 | 13 | 2027 | AAH46457.1 |

Genomic Map