Assay Details
Target Gene Details
Entrez Gene ID: | 65960 |
Gene Name: | twisted gastrulation homolog 1 (Drosophila) |
Gene Aliases: |
1810013J15Rik, 9030422N06Rik, AW552143, D17Ertd403e, Tsg, Twg |
Location: |
Chr.17:65923065-65951187 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Twsg1 | NM_023053.3 | 4 | 429 | NP_075540.1 |
AF295097.1 | 3 | 287 | AAG10080.1 | |
AJ297390.1 | 4 | 379 | CAC05586.1 | |
AK033504.1 | 4 | 388 | BAC28326.1 | |
AK049646.1 | 4 | 423 | BAC33857.1 | |
AK168547.1 | 4 | 379 | BAE40422.1 | |
AK168633.1 | 4 | 388 | BAE40493.1 | |
AK191967.1 | ||||
BC004850.1 | 4 | 384 | AAH04850.1 | |
CJ165542.1 |
