Assay Details
Target Gene Details
Entrez Gene ID: | 81500 |
Gene Name: | endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae) |
Gene Aliases: |
1810057E01Rik, AI042831, wz |
Location: |
Chr.18:35266396-35499052 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sil1 | NM_030749.2 | NP_109674.2 | ||
| AJ297884.1 | CAC17789.1 | |||
| AK170415.1 | BAE41781.1 | |||
| BC016119.1 | AAH16119.1 | |||
| BC016466.1 | AAH16466.1 | |||
| CX219879.1 |
More Information
Set Membership: |
|

Genomic Map