Assay Details
Target Gene Details
Entrez Gene ID: | 83965 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 5 |
Gene Aliases: |
AW061004, D17Abb1e, E-NPP 5, mKIAA4152 |
Location: |
Chr.17:44078796-44086567 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp5 | NM_001168620.1 | 2 | 478 | NP_001162091.1 |
NM_032003.2 | 2 | 564 | NP_114392.1 | |
AF233377.1 | 2 | 541 | AAG49143.1 | |
AK032341.1 | 2 | 467 | ||
AK210356.1 | ||||
AK218326.1 | ||||
AK220510.1 | 2 | 507 | BAD90517.1 | |
BC089320.1 | 3 | 284 | AAH89320.1 | |
BC138977.1 | 2 | 479 | ||
BC138978.1 | 2 | 479 |
