Assay Details
Target Gene Details
Entrez Gene ID: | 83965 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 5 |
Gene Aliases: |
AW061004, D17Abb1e, E-NPP 5, mKIAA4152 |
Location: |
Chr.17:44078796-44086567 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpp5 | NM_001168620.1 | 4 | 1544 | NP_001162091.1 |
| NM_032003.2 | 4 | 1630 | NP_114392.1 | |
| AF233377.1 | 4 | 1607 | AAG49143.1 | |
| AK032341.1 | 4 | 1533 | ||
| AK220510.1 | 4 | 1573 | BAD90517.1 | |
| BC089320.1 | 4 | 3634 | AAH89320.1 | |
| BC138977.1 | 4 | 1545 | ||
| BC138978.1 | 4 | 1545 |

Genomic Map