Assay Details
Target Gene Details
Entrez Gene ID: | 66530 |
Gene Name: | UBX domain protein 6 |
Gene Aliases: |
1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2 |
Location: |
Chr.17:56068249-56074989 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubxn6 | NM_024432.2 | 11 | 1416 | NP_077752.1 |
| AF272895.1 | AAK13259.1 | |||
| AK008942.1 | 11 | 1424 | ||
| AK009117.1 | 10 | 1198 | BAB26082.1 | |
| AK075705.1 | 9 | 2480 | ||
| AK131187.1 | 5 | 3188 | BAD21437.1 | |
| AK161273.1 | 11 | 1438 | BAE36284.1 | |
| AK220061.1 | 1 | 103 | ||
| BC016412.1 | AAH16412.1 | |||
| BC049963.1 | 11 | 1416 | AAH49963.1 |

Genomic Map