Assay Details
Target Gene Details
Entrez Gene ID: | 67064 |
Gene Name: | charged multivesicular body protein 1B |
Gene Aliases: |
2810405I11Rik, Chmp1b1 |
Location: |
Chr.18:67205359-67207887 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp1b | NM_024190.2 | 1 | 587 | NP_077152.1 |
| AK012996.1 | 1 | 537 | ||
| AK014091.1 | 1 | 554 | BAB29150.2 | |
| AK151071.1 | 1 | 556 | BAE30087.1 | |
| AK167614.1 | 1 | 526 | BAE39667.1 | |
| BC002229.1 | 1 | 523 | AAH02229.1 | |
| CT010307.1 | 1 | 445 | CAJ18515.1 |
Target Gene Details
Entrez Gene ID: | 14680 |
Gene Name: | guanine nucleotide binding protein, alpha stimulating, olfactory type |
Gene Aliases: |
2610011C15Rik, 9630020G10Rik, Galphaolf, Gna10, Golf |
Location: |
Chr.18:67088298-67226792 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnal | NM_010307.3 | NP_034437.1 | ||
| NM_177137.5 | NP_796111.2 | |||
| AK158831.1 | BAE34685.1 | |||
| AY179169.1 | AAO03564.1 | |||
| BC078439.1 | AAH78439.1 |
More Information
Set Membership: |
|

Genomic Map