Assay Details
Target Gene Details
Entrez Gene ID: | 224105 |
Gene Name: | p21 protein (Cdc42/Rac)-activated kinase 2 |
Gene Aliases: |
5330420P17Rik, A130002K10Rik, AI836325, D16Ertd269e, PAK-2, mKIAA4182 |
Location: |
Chr.16:32016290-32079342 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak2 | NM_177326.3 | 15 | 3368 | NP_796300.1 |
| AK037417.1 | BAC29802.1 | |||
| AK038324.1 | BAC29970.1 | |||
| AK076227.1 | 3 | 1845 | BAC36261.1 | |
| AK152023.1 | 1 | 762 | ||
| AK220528.1 | 6 | 2197 | BAD90309.1 | |
| BC029195.1 | 4 | 1959 | AAH29195.1 | |
| BC086650.1 | 15 | 3299 | AAH86650.1 |
More Information
Set Membership: |
|

Genomic Map