Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm37013 | NM_001174154.2 | 4 | 2970 | NP_001167625.1 |
AF464161.1 | ||||
AK189050.1 | ||||
AK194976.1 | ||||
AK196255.1 | ||||
AK216085.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm38666 | NM_001301256.1 | 4 | 2974 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha7-g | NM_001301259.1 | 4 | 2944 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 93709 |
Gene Name: | protocadherin gamma subfamily A, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37661945-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga1 | NM_033584.1 | 4 | 2888 | NP_291062.1 |
AY013792.1 | 4 | 2888 | AAK26081.1 |
Target Gene Details
Entrez Gene ID: | 93722 |
Gene Name: | protocadherin gamma subfamily A, 10 |
Gene Aliases: |
- |
Location: |
Chr.18:37747188-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga10 | NM_033593.3 | 4 | 2885 | NP_291071.1 |
AY013793.1 | 4 | 2885 | AAK26082.1 | |
BC035540.1 | 4 | 3040 | AAH35540.1 |
Target Gene Details
Entrez Gene ID: | 93723 |
Gene Name: | protocadherin gamma subfamily A, 11 |
Gene Aliases: |
- |
Location: |
Chr.18:37755773-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga11 | NM_033594.2 | 4 | 3062 | NP_291072.1 |
AY013794.1 | 4 | 2894 | AAK26083.1 | |
BC059837.1 | 4 | 2993 | AAH59837.1 | |
BC064675.1 | 4 | 3062 | AAH64675.1 |
Target Gene Details
Entrez Gene ID: | 93724 |
Gene Name: | protocadherin gamma subfamily A, 12 |
Gene Aliases: |
Pcdh13, pc2c |
Location: |
Chr.18:37765580-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga12 | NM_033595.4 | 4 | 3428 | NP_291073.1 |
AF464156.1 | 4 | 510 | AAM93559.1 | |
AY013795.1 | 4 | 2891 | AAK26084.1 | |
BC054528.1 | 4 | 3019 | AAH54528.1 | |
BC056998.1 | 4 | 3428 | AAH56998.1 | |
U88550.1 | 4 | 2984 | AAD00652.1 |
Target Gene Details
Entrez Gene ID: | 93710 |
Gene Name: | protocadherin gamma subfamily A, 2 |
Gene Aliases: |
mKIAA0588 |
Location: |
Chr.18:37669105-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga2 | NM_033585.1 | 4 | 2888 | NP_291063.1 |
AB093250.1 | 4 | 3040 | BAC41434.1 | |
AY013796.1 | 4 | 2888 | AAK26085.1 | |
BC139178.1 | 4 | 2957 |
Target Gene Details
Entrez Gene ID: | 93711 |
Gene Name: | protocadherin gamma subfamily A, 3 |
Gene Aliases: |
- |
Location: |
Chr.18:37674335-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga3 | NM_033586.2 | 4 | 3040 | NP_291064.1 |
AY013797.1 | 4 | 2879 | AAK26086.1 | |
BC060691.1 | 4 | 3040 | AAH60691.1 |
Target Gene Details
Entrez Gene ID: | 93712 |
Gene Name: | protocadherin gamma subfamily A, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37685400-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga4 | NM_033587.3 | 4 | 2885 | NP_291065.3 |
AY013798.1 | 4 | 2885 | AAK26087.1 | |
BC049814.1 | 4 | 3011 | AAH49814.1 |
Target Gene Details
Entrez Gene ID: | 93713 |
Gene Name: | protocadherin gamma subfamily A, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37694501-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga5 | NM_033588.4 | 4 | 2891 | NP_291066.1 |
AF464162.1 | ||||
AK043491.1 | 4 | 2968 | BAC31558.1 | |
AY013799.1 | 4 | 2891 | AAK26088.1 | |
BC057404.1 | 4 | 2991 | AAH57404.1 |
Target Gene Details
Entrez Gene ID: | 93714 |
Gene Name: | protocadherin gamma subfamily A, 6 |
Gene Aliases: |
- |
Location: |
Chr.18:37707229-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga6 | NM_033589.1 | 4 | 2891 | NP_291067.1 |
AY013800.1 | 4 | 2891 | AAK26089.1 | |
BC080852.1 | 4 | 2936 | AAH80852.1 |
Target Gene Details
Entrez Gene ID: | 93715 |
Gene Name: | protocadherin gamma subfamily A, 7 |
Gene Aliases: |
- |
Location: |
Chr.18:37714834-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga7 | NM_033590.3 | 4 | 3008 | NP_291068.2 |
AY013801.1 | 4 | 2900 | AAK26090.1 | |
BC077712.1 | 4 | 3008 | AAH77712.1 |
Target Gene Details
Entrez Gene ID: | 93716 |
Gene Name: | protocadherin gamma subfamily A, 8 |
Gene Aliases: |
mKIAA4054 |
Location: |
Chr.18:37725706-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga8 | NM_033591.3 | 4 | 3078 | NP_291069.1 |
AK220461.1 | 4 | 3031 | BAD90279.1 | |
AY013802.1 | 4 | 2891 | AAK26091.1 | |
BC054555.1 | 4 | 3039 | AAH54555.1 | |
BC054787.1 | 4 | 3078 | AAH54787.1 |
Target Gene Details
Entrez Gene ID: | 93717 |
Gene Name: | protocadherin gamma subfamily A, 9 |
Gene Aliases: |
- |
Location: |
Chr.18:37736936-37841863 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga9 | NM_033592.3 | 4 | 3075 | NP_291070.1 |
AY013803.1 | 4 | 2891 | AAK26092.1 | |
BC079567.1 | 4 | 3075 | AAH79567.1 |
Target Gene Details
Entrez Gene ID: | 93699 |
Gene Name: | protocadherin gamma subfamily B, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37680458-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb1 | NM_033574.3 | 4 | 2849 | NP_291052.3 |
AY013804.1 | 4 | 2849 | AAK26093.1 | |
BC054741.1 | 4 | 3016 | AAH54741.1 | |
BC098367.1 | 5 | 2655 | AAH98367.1 |
Target Gene Details
Entrez Gene ID: | 93700 |
Gene Name: | protocadherin gamma subfamily B, 2 |
Gene Aliases: |
- |
Location: |
Chr.18:37689859-37841872 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb2 | NM_033575.3 | 4 | 2978 | NP_291053.1 |
AF464151.1 | 4 | 619 | AAM93554.1 | |
AY013805.1 | 4 | 2879 | AAK26094.1 | |
BC054557.1 | 4 | 2978 | AAH54557.1 |
Target Gene Details
Entrez Gene ID: | 93701 |
Gene Name: | protocadherin gamma subfamily B, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37720554-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb4 | NM_033576.1 | 4 | 2831 | NP_291054.1 |
AF464164.1 | AAM93565.1 | |||
AY013806.1 | 4 | 2831 | AAK26095.1 | |
BC138701.1 | 4 | 2927 |
Target Gene Details
Entrez Gene ID: | 93702 |
Gene Name: | protocadherin gamma subfamily B, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37731154-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb5 | NM_033577.1 | 4 | 2873 | NP_291055.1 |
AY013807.1 | 4 | 2873 | AAK26096.1 |
Target Gene Details
Entrez Gene ID: | 93703 |
Gene Name: | protocadherin gamma subfamily B, 6 |
Gene Aliases: |
- |
Location: |
Chr.18:37742094-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb6 | NM_033578.3 | 4 | 3032 | NP_291056.1 |
AY013808.1 | 4 | 2885 | AAK26097.1 | |
BC054556.1 | 4 | 3032 | AAH54556.1 |
Target Gene Details
Entrez Gene ID: | 93704 |
Gene Name: | protocadherin gamma subfamily B, 7 |
Gene Aliases: |
- |
Location: |
Chr.18:37751779-37841870 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb7 | NM_033579.1 | 4 | 2882 | NP_291057.1 |
AY013809.1 | 4 | 2882 | AAK26098.1 | |
BC066823.1 | 4 | 3036 | AAH66823.1 |
Target Gene Details
Entrez Gene ID: | 93705 |
Gene Name: | protocadherin gamma subfamily B, 8 |
Gene Aliases: |
- |
Location: |
Chr.18:37761801-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb8 | NM_033580.2 | 4 | 2966 | NP_291058.1 |
AY013810.1 | 4 | 2888 | AAK26099.1 | |
BC139099.1 | 4 | 2966 | ||
BC139100.1 | 4 | 2966 |
Target Gene Details
Entrez Gene ID: | 93706 |
Gene Name: | protocadherin gamma subfamily C, 3 |
Gene Aliases: |
PC43, Pcdh2 |
Location: |
Chr.18:37806410-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgc3 | NM_033581.3 | 4 | 3035 | NP_291059.1 |
AK086361.1 | 4 | 2669 | ||
AY013811.1 | 4 | 2897 | AAK26100.1 | |
BC059175.1 | 4 | 3035 | AAH59175.1 | |
BC098457.1 | 5 | 1937 | AAH98457.1 |
Target Gene Details
Entrez Gene ID: | 93707 |
Gene Name: | protocadherin gamma subfamily C, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37815079-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgc4 | NM_033582.2 | 4 | 3372 | NP_291060.1 |
AK139349.1 | 4 | 2110 | BAE23971.1 | |
AY013812.1 | 4 | 2918 | AAK26101.1 | |
BC064808.1 | 4 | 3372 | AAH64808.1 |
Target Gene Details
Entrez Gene ID: | 93708 |
Gene Name: | protocadherin gamma subfamily C, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37819546-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgc5 | NM_033583.3 | 4 | 3056 | NP_291061.1 |
AF464163.1 | 4 | 3056 | AAM93564.1 | |
AY013813.1 | 4 | 2927 | AAK26102.1 | |
BC138677.1 | 4 | 3000 | ||
BC138678.1 | 4 | 3114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
