Assay Details
Target Gene Details
Entrez Gene ID: | 224814 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 10 |
Gene Aliases: |
Mrp7, mFLJ00002 |
Location: |
Chr.17:46303230-46328032 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcc10 | NM_170680.2 | 1 | 207 | NP_733780.1 |
| AF417121.1 | 1 | 188 | AAM18536.1 | |
| AK131108.1 | 1 | 114 | BAD21358.1 | |
| AK133240.1 | 1 | 164 | BAE21573.1 | |
| AK164902.1 | 1 | 181 | ||
| AK170539.1 | 1 | 181 | ||
| AK170600.1 | 1 | 181 | BAE41905.1 | |
| AK171535.1 | 1 | 182 | ||
| BC150818.1 | 1 | 160 |
More Information
Set Membership: |
|

Genomic Map