Assay Details
Target Gene Details
Entrez Gene ID: | 381110 |
Gene Name: | regulator of microtubule dynamics 2 |
Gene Aliases: |
AW061290, Fam82a1, RMD-2, mRMD-2 |
Location: |
Chr.17:79578598-79685067 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rmdn2 | NM_201361.2 | 2 | 464 | NP_958749.1 |
AK034617.1 | 2 | 466 | BAC28771.1 | |
AK139382.1 | 2 | 683 | BAE23985.1 | |
BC024059.1 | 2 | 544 | AAH24059.1 |
