Assay Details
Target Gene Details
Entrez Gene ID: | 72238 |
Gene Name: | TBC1 domain family, member 5 |
Gene Aliases: |
1600014N05Rik |
Location: |
Chr.17:50733124-51180198 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d5 | NM_001285991.1 | 17 | 1754 | NP_001272920.1 |
| NM_001285993.1 | 17 | 1721 | NP_001272922.1 | |
| NM_028162.4 | 17 | 1754 | NP_082438.3 | |
| AK144467.1 | 17 | 1755 | BAE25904.1 | |
| AK154975.1 | 17 | 1714 | BAE32965.1 | |
| AK163366.1 | 17 | 1783 | ||
| AK220251.1 | 17 | 1721 | BAD90176.1 | |
| BC043113.1 | 17 | 1570 | AAH43113.1 | |
| BC098328.1 | 12 | 1100 | AAH98328.1 |
More Information
Set Membership: |
|

Genomic Map