Assay Details
Target Gene Details
Entrez Gene ID: | 21425 |
Gene Name: | transcription factor EB |
Gene Aliases: |
Tcfeb, bHLHe35 |
Location: |
Chr.17:47737037-47792416 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfeb | NM_001161722.1 | 1 | 233 | NP_001155194.1 |
AK078755.1 | 1 | 165 | BAC37377.1 |
