Assay Details
Target Gene Details
Entrez Gene ID: | 70573 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
5730478M09Rik, AU015083, BB165076 |
Location: |
Chr.16:22813215-22857701 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbccd1 | NM_001081368.1 | 6 | 1651 | NP_001074837.1 |
AK039905.1 | 6 | 1476 | BAC30476.1 | |
AK040787.1 | 7 | 1704 | ||
AK133804.1 | 6 | 1592 | ||
AK188790.1 | ||||
AK211627.1 | ||||
AK211897.1 | 1 | 184 | ||
BC079629.1 | 1 | 258 | AAH79629.1 | |
BC082558.1 | 6 | 1620 | AAH82558.1 | |
DQ547931.1 |
