Assay Details
Target Gene Details
Entrez Gene ID: | 70573 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
5730478M09Rik, AU015083, BB165076 |
Location: |
Chr.16:22813215-22857701 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbccd1 | NM_001081368.1 | 8 | 2455 | NP_001074837.1 |
AK039905.1 | 8 | 2280 | BAC30476.1 | |
AK133804.1 | 8 | 2396 | ||
BC079629.1 | 3 | 1062 | AAH79629.1 | |
BC082558.1 | 8 | 2424 | AAH82558.1 |
