Assay Details
Target Gene Details
Entrez Gene ID: | 14913 |
Gene Name: | guanylate cyclase activator 1a (retina) |
Gene Aliases: |
GC-A, Gcap1, Guca1, mGCAP1 |
Location: |
Chr.17:47394558-47413514 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Guca1a | NM_008189.2 | 1 | 252 | NP_032215.2 |
BC026834.1 | 1 | 252 | AAH26834.1 | |
BC031810.1 | 1 | 219 | AAH31810.1 | |
L36860.1 | 1 | 109 | AAA60716.1 |
