Assay Details
Target Gene Details
Entrez Gene ID: | 23984 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
- |
Location: |
Chr.17:8525187-8986648 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde10a | NM_001290707.1 | NP_001277636.1 | ||
| NM_011866.2 | 1 | 202 | NP_035996.2 | |
| AK014090.1 | 1 | 255 | ||
| AK039249.1 | BAC30292.1 | |||
| AK046535.1 | ||||
| AK147690.1 | 1 | 169 | ||
| AY360383.1 | 1 | 202 | AAR12579.1 |
More Information
Set Membership: |
|

Genomic Map