Assay Details
Target Gene Details
Entrez Gene ID: | 18701 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class F |
Gene Aliases: |
- |
Location: |
Chr.17:86997258-87025471 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigf | NM_008838.1 | NP_032864.1 | ||
| AK132796.1 | BAE21363.1 | |||
| AK171083.1 | BAE42236.1 | |||
| BC028862.1 | AAH28862.1 | |||
| D50264.1 | BAA08818.1 |
Target Gene Details
Entrez Gene ID: | 104215 |
Gene Name: | ras homolog gene family, member Q |
Gene Aliases: |
Arhq, TC10A, Tc10 |
Location: |
Chr.17:86963111-87000069 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhoq | NM_145491.2 | 5 | 3060 | NP_663466.2 |
| AB060650.1 | 5 | 2878 | BAB91068.1 | |
| BC048813.1 | AAH48813.2 | |||
| BC056363.1 | 5 | 3060 | AAH56363.1 |
More Information
Set Membership: |
|

Genomic Map