Assay Details
Target Gene Details
Entrez Gene ID: | 21425 |
Gene Name: | transcription factor EB |
Gene Aliases: |
Tcfeb, bHLHe35 |
Location: |
Chr.17:47737037-47792416 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfeb | NM_001161722.1 | 3 | 549 | NP_001155194.1 |
NM_001161723.1 | 3 | 543 | NP_001155195.1 | |
NM_011549.3 | 2 | 490 | NP_035679.3 | |
AF079095.1 | 2 | 471 | AAD20979.1 | |
AK078755.1 | 3 | 481 | BAC37377.1 | |
AK146016.1 | 2 | 515 | BAE26834.1 | |
AK154971.1 | 3 | 547 | BAE32963.1 | |
BC064789.1 | 2 | 490 | AAH64789.1 |
