Assay Details
Target Gene Details
Entrez Gene ID: | 269033 |
Gene Name: | RIKEN cDNA 4930503L19 gene |
Gene Aliases: |
9930107F24, BB148262, Lars, Las2 |
Location: |
Chr.18:70452210-70472504 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930503L19Rik | NM_172967.2 | 2 | 341 | NP_766555.1 |
AK037066.1 | 2 | 342 | BAC29691.1 | |
AK080780.1 | 2 | 371 | BAC38019.1 | |
AK087788.1 | 2 | 314 | BAC40002.1 | |
AK148410.1 | 2 | 365 | BAE28537.1 | |
AK160248.1 | 2 | 216 | ||
AK160714.1 | 2 | 311 | ||
BC057927.1 | 1 | 58 | AAH57927.1 |
